Merck
  • Effect of water hardness on the toxicity of cobalt and nickel to a freshwater fish, Capoeta fusca.
Effect of water hardness on the toxicity of cobalt and nickel to a freshwater fish, Capoeta fusca.
Biomedical and environmental sciences : BES (2012-03-01)
Alireza Pourkhabbaz, Tahereh Khazaei, Samira Behravesh, Mohammad Ebrahimpour, Hamidreza Pourkhabbaz
ZUSAMMENFASSUNG
To determine the effects of water hardness on the toxicities of cobalt (Co) and nickel (Ni) to a freshwater fish, Capoeta fusca. Toxicity was investigated by static bioassay. Fish were exposed to cobalt (as CoCl(2)) and nickel (as NiCl(2)) for 96 h in waters with two levels of hardness ("hard" and "very hard", nominally 130 mg/L and 350 mg/L as CaCO(3), respectively). Water hardness had a significant effect on the acute toxicity of both elements. The 96 h LC(50) values for Co were 91.7 mg/L and 204.8 mg/L in hard and very hard waters, respectively, and for Ni the 96 h LC(50) values were 78.0 mg/L and 127.2 mg/L, respectively. The fish were more sensitive to Co and Ni toxicity in hard water than in very hard water; very hard water protects C. fusca against the toxicity of Co and Ni.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, ReagentPlus®
Sigma-Aldrich
Nickel(II)-chlorid, 98%
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, 99.9% trace metals basis
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, puriss. p.a., ≥98%
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, BioReagent, suitable for cell culture
Sigma-Aldrich
Nickel(II)-chlorid, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Nickel(II)-chlorid Hydrat, 99.95% trace metals basis
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, 99.999% trace metals basis