Merck
  • Effect of carbon sources on the removal of 1,1,2-trichloroethane and 1,1,2,2-tetrachloroethane in UASB reactor.
Effect of carbon sources on the removal of 1,1,2-trichloroethane and 1,1,2,2-tetrachloroethane in UASB reactor.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2012-03-02)
Debolina Basu, Shyam R Asolekar
ZUSAMMENFASSUNG
The effect of two carbon sources namely sodium acetate and ethanol was studied in bench-scale Upflow Anaerobic Sludge Blanket (UASB) reactors for the removal of chlorinated ethanes i.e., 1,1,2-Trichloroethane (TCA) and 1,1,2,2-Tetrachloroethane (TeCA) contained in the simulated wastewaters. The Hydraulic Retention Time (HRT) was maintained as 24 hours in all the reactors. The granular biomass in the test reactors R2 and R3 were acclimated to 40 mg/L of TCA and 20 mg/L of TeCA, respectively. The effluent TCA and TeCA concentrations were 0.03 mg/L and 0.18 mg/L, respectively, at the end of acclimation phase. Sodium acetate and ethanol both were found to be suitable as the primary substrates in the biodegradation of TCA and TeCA. However, lower concentrations of the toxic pollutants (TCA and TeCA) were obtained in the effluents with the use of sodium acetate. The COD removal efficiency in the test reactors (R2 and R3) varied in the range of 95 % to 98.2 % accompanied by the formation of 1,2-Dichloroethane (DCA) as the major intermediate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1,2,2-Tetrachlorethan, reagent grade, ≥98%
Sigma-Aldrich
1,1,2-Trichlorethan, contains ≤3% 2-propanol as stabilizer, 97%
Supelco
1,1,2,2-Tetrachlorethan, analytical standard
Supelco
1,1,2-Trichlorethan, analytical standard