Merck
Synthesis of functional acetylene derivatives from calcium carbide.
ChemSusChem (2012-03-02)
Zhewang Lin, Dingyi Yu, Yin Ngai Sum, Yugen Zhang
ZUSAMMENFASSUNG
AHA Erlebnis: CaC(2), used to produce acetylene until several decades ago, is re-emerging as a cheap, sustainable resource synthesized from coal and lignocellulosic biomass. We report efficient catalytic protocols for the synthesis of functional acetylene derivatives from CaC(2) through aldehyde, alkyne, and amine (AAA) as well as alkyne, haloalkane, and amine (AHA) couplings, and in addition demonstrate its use in click and Sonogashira chemistry, showing that calcium carbide is a sustainable and cost-efficient carbon source.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calciumcarbid, granulated, technical, ≥75% (gas-volumetric)
Sigma-Aldrich
Calciumcarbid, pieces, thickness <10 mm , typically, technical grade, ~80%