Merck
  • Molecular drug design, synthesis and structure elucidation of a new specific target peptide based metallo drug for cancer chemotherapy as topoisomerase I inhibitor.
Molecular drug design, synthesis and structure elucidation of a new specific target peptide based metallo drug for cancer chemotherapy as topoisomerase I inhibitor.
Dalton transactions (Cambridge, England : 2003) (2012-03-13)
Sartaj Tabassum, Waddhaah M Al-Asbahy, Mohd Afzal, Farukh Arjmand, Vivek Bagchi
ZUSAMMENFASSUNG
To evaluate the biological preference of metallopeptide drugs in cancer cells, a new dinuclear copper(II) complex [Cu(2)(glygly)(2)(ppz)(H(2)O)(4)]·2H(2)O (1) (glygly = glycyl glycine anion and ppz = piperazine), was designed and synthesized as topoisomerase I inhibitor. The structural elucidation of the complex was done by elemental analysis, spectroscopic methods and single crystal X-ray diffraction. The in vitro DNA binding studies of complex 1 with CT DNA were carried out by employing different optical methods viz. UV-vis, fluorescence and circular dichroism. The molecular docking technique was also utilized to ascertain the mechanism and mode of action towards the molecular target DNA and enzymes. Complex 1 cleaves pBR322 DNA via an oxidative mechanism and strongly binds to the DNA minor groove. Furthermore, complex 1 exhibits significant inhibitory effects on the catalytic activity of topoisomerase I at a very low concentration, ~12.5 μM, in addition to its excellent SOD mimics (IC(50)~0.086 μM).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
Nitrotetrazoliumblauchlorid, tablet
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, powder, electrophoresis grade
Sigma-Aldrich
Gly-Gly, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Sigma-Aldrich
Gly-Gly, BioUltra, ≥99.5% (NT)