Merck
  • A SAR study of novel antiproliferative ruthenium and osmium complexes with quinoxalinone ligands in human cancer cell lines.
A SAR study of novel antiproliferative ruthenium and osmium complexes with quinoxalinone ligands in human cancer cell lines.
Journal of medicinal chemistry (2012-03-16)
Werner Ginzinger, Gerhard Mühlgassner, Vladimir B Arion, Michael A Jakupec, Alexander Roller, Markus Galanski, Michael Reithofer, Walter Berger, Bernhard K Keppler
ZUSAMMENFASSUNG
A series of ruthenium(II) arene complexes with 3-(1H-benzimidazol-2-yl)-1H-quinoxalin-2-one, bearing pharmacophoric groups of known protein kinase inhibitors, and related benzoxazole and benzothiazole derivatives have been synthesized. In addition, the corresponding osmium complexes of the unsubstituted ligands have also been prepared. The compounds have been characterized by NMR, UV-vis, and IR spectroscopy, ESI mass spectrometry, elemental analysis, and by X-ray crystallography. Antiproliferative activity in three human cancer cell lines (A549, CH1, SW480) was determined by MTT assays, yielding IC(50) values of 6-60 μM for three unsubstituted metal-free ligands, whereas values for the metal complexes vary in a broad range from 0.3 to 140 μM. Complexation with osmium of quinoxalinone derivatives with benzimidazole or benzothiazole results in a more consistent increase in cytotoxicity than complexation with ruthenium. For selected compounds, the capacity to induce apoptosis was confirmed by fluorescence microscopy and flow-cytometric analysis, whereas cell cycle effects are only moderate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Osmium, powder, 99.9% trace metals basis