Merck
  • Microencapsulation of menhaden fish oil containing soluble rice bran fiber using spray drying technology.
Microencapsulation of menhaden fish oil containing soluble rice bran fiber using spray drying technology.
Journal of food science (2012-03-16)
Yuting Wan, Joseph David Bankston, Peter J Bechtel, Subramaniam Sathivel
ZUSAMMENFASSUNG
Emulsion (EFMO) containing purified menhaden oil (PMO) and soluble rice bran fiber (SRBF) was dried in a pilot scale spray dryer and produced microencapsulated PMO with SRBF (MFMO). EFMO had well isolated spherical droplets with the size of 1 to 10 μm and showed pseudoplastic fluid and viscoelastic characteristics. EFMO had lower lipid oxidation than the emulsion containing PMO without SRBF when both emulsions were stored at 20 and 40 °C for 88 h, which indicated that the SRBF reduced the lipid oxidation in the EFMO. The estimated MFMO production rate (3.45 × 10(-5) kg dry solids/s) was higher than the actual production rate (2.31 × 10(-5) kg dry solids/s). The energy required to spray dry the EFMO was 12232 kJ/kg of emulsion. EPA and DHA contents of MFMO were 11.52% and 4.51%, respectively. The particle size of 90% MFMO ranged from 8 to 62 μm, and the volume-length diameter of MFMO was 28.5 μm.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fischöl aus Menhaden, crude source of omega-3 fatty acids