Merck
Tetravalent dysprosium in a perovskite-type oxide.
Advanced materials (Deerfield Beach, Fla.) (2012-03-21)
Donglin Han, Tetsuya Uda, Yoshitaro Nose, Toshihiro Okajima, Hidenobu Murata, Isao Tanaka, Kozo Shinoda
ZUSAMMENFASSUNG
The existence of tetravalent dysprosium in perovskite-type oxide barium zirconate is confirmed in this work. This discovery will stimulate many researchers in diverse fields such as catalysts, solid state ionics, sensors, and fluorescent materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bariumoxid, 99.99% trace metals basis
Sigma-Aldrich
Bariumoxid, 97%
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis