Merck
Hypolipidemic effect of diet supplementation with bacterial levan in cholesterol-fed rats.
International journal of biological macromolecules (2012-03-22)
Karima Srih Belghith, Imen Dahech, Khaled Hamden, Abdelfattah Feki, Hafedh Mejdoub, Hafedh Belghith
ZUSAMMENFASSUNG
Levan polysaccharide, a type of fructan, has been shown to have industrial applications as a new industrial gum in the fields of cosmetics, foods like dietary fiber and pharmaceutical goods. The objective of this current study was to investigate the possible hypolipidemic and antioxidative effects of levan in rats fed with a high-cholesterol diet. Animals were allocated into four groups of six rats each: a normal diet group (Control), normal rats received levan (L), a high-cholesterol diet group (Chol) and a high-cholesterol diet with a daily dose of levan equivalent to 5%. Treated hypercholesterolemic rats were administrated with levan in drinking water through oral gavage for 60 days. After the treatment period, the plasma antioxidant enzymes and lipid profiles were determined. Our results show that treatment with levan polysaccharide positively changed plasma antioxidant enzyme activities and lipid profiles (total cholesterol, HDL-cholesterol, LDL-cholesterol and triglycerides) in cholesterol-rats, and thus may have potential hypolipidemic and antioxidant effects. Levan could protect against oxidative stress linked atherosclerosis and decrease the atherogenic index.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Levan aus Erwinia herbicola, ≥98% (TLC)