Merck
Close-packed monolayers of charged Janus-type nanoparticles at the air-water interface.
Journal of colloid and interface science (2012-03-24)
Volodymyr Sashuk, Robert Hołyst, Tomasz Wojciechowski, Marcin Fiałkowski
ZUSAMMENFASSUNG
We present a new method to obtain close-packed monolayers composed of noble metal nanoparticles (NP) possessing well-defined permanent charge of either sign. The method is based on the fact that ligands forming the protecting layer exhibit ability to rearrange at the NPs surface. We demonstrate that if the protecting layer is composed of a mixture of hydrophobic and hydrophilic charged ligands in appropriate proportion, the NP exhibits properties of a Janus-type particle with one of the hemispheres hydrophilic and the other hydrophobic. Such amphiphilic NPs self-assemble into a monolayer of well defined surface charge at the air-water interface. Due to strong stabilizing effect of the lateral electrostatic repulsions, such monolayer can be compressed to form close-packed hexagonal structure, and then easily transferred onto a solid substrate with the Langmuir-Blodgett technique.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Caprinsäure, ≥98.0%
Sigma-Aldrich
Caprinsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Natrium-caprinat, ≥98%
Sigma-Aldrich
Dodecylamin, ≥99%
Sigma-Aldrich
Dodecylamin, 98%
Sigma-Aldrich
Caprinsäure, natural, ≥98%, FCC, FG
Supelco
Caprinsäure, analytical standard
Sigma-Aldrich
Dodecylamin, puriss., ≥99.5% (GC)