Merck
  • A homochiral luminescent 2D porous coordination polymer with collagen-type triple helices showing selective guest inclusion.
A homochiral luminescent 2D porous coordination polymer with collagen-type triple helices showing selective guest inclusion.
Inorganic chemistry (2012-03-28)
Biplab Joarder, Abhijeet K Chaudhari, Sujit K Ghosh
ZUSAMMENFASSUNG
A homochiral luminescent porous coordination polymer, [Cd(L)(H(2)O)]·3H(2)O, with interconnected collagen like triple-helical chains has been synthesized solvothermally by using cadmium(II) salt and a newly designed d-isosorbide-based, enantiomerically pure chiral ligand. The framework is a 2D porous material and forms a 1D channel along the a axis, with the channel dimensions ~6.2 × 4.4 Å(2). The compound has high selectivity in the uptake of water and methanol over other solvents (e.g., tetrahydrofuran, ethanol, benzene, and cyclohexane) inside the channels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dianhydro-D-glucit, 98%