Merck
[Survey of plasticizers in polyvinyl chloride toys].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2012-03-28)
Yutaka Abe, Miku Yamaguchi, Motoh Mutsuga, Yoshichika Hirahara, Yoko Kawamura
ZUSAMMENFASSUNG
Plasticizers in 101 samples of polyvinyl chloride (PVC) toys on the Japanese market were surveyed. No phthalates were detected in designated toys, though bis(2-ethylhexyl)phthalate, diisononyl phthalate, diisobutyl phthalate, dibutyl phthalate, diisodecyl phthalate and benzyl butyl phthalate were detected in more than half of other toys. 2,2,4-Tributyl-1,3-pentanediol diisobutylate, o-acetyl tributyl citrate, adipates and diacetyl lauroyl glycerol, which are alternative plasticizers to phthalates, were detected. The results of structural analysis confirmed the presence of di(2-ethylhexyl)terephthalate, tributyl citrate, diisononyl 1,2-cyclohexanedicarboxylate and neopentyl glycol esters; these have not previonsly been reported in Japan. There appears to be a shift in plasticizers used for designated toys from phthalates to new plasticizers, and the number of different plasticizers is increasing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Terephthalsäure, 98%
Sigma-Aldrich
Tributylcitrat, ≥97.0%