Merck
  • Protein kinase A enhances lipopolysaccharide-induced IL-6, IL-8, and PGE₂ production by human gingival fibroblasts.
Protein kinase A enhances lipopolysaccharide-induced IL-6, IL-8, and PGE₂ production by human gingival fibroblasts.
Journal of negative results in biomedicine (2012-03-29)
Toshiaki Ara, Yoshiaki Fujinami, Hiroko Urano, Kaname Hirai, Toshimi Hatori, Hiroo Miyazawa
ZUSAMMENFASSUNG
Periodontal disease is accompanied by inflammation of the gingiva and destruction of periodontal tissues, leading to alveolar bone loss in severe clinical cases. Interleukin (IL)-6, IL-8, and the chemical mediator prostaglandin E₂ (PGE₂) are known to play important roles in inflammatory responses and tissue degradation. Recently, we reported that the protein kinase A (PKA) inhibitor H-89 suppresses lipopolysaccharide (LPS)-induced IL-8 production by human gingival fibroblasts (HGFs). In the present study, the relevance of the PKA activity and two PKA-activating drugs, aminophylline and adrenaline, to LPS-induced inflammatory cytokines (IL-6 and IL-8) and PGE₂ by HGFs were examined. HGFs were treated with LPS from Porphyromonas gingivalis and H-89, the cAMP analog dibutyryl cyclic AMP (dbcAMP), aminophylline, or adrenaline. After 24 h, IL-6, IL-8, and PGE₂ levels were evaluated by ELISA. H-89 did not affect LPS-induced IL-6 production, but suppressed IL-8 and PGE₂ production. In contrast, dbcAMP significantly increased LPS-induced IL-6, IL-8, and PGE₂ production. Up to 10 μg/ml of aminophylline did not affect LPS-induced IL-6, IL-8, or PGE₂ production, but they were significantly increased at 100 μg/ml. Similarly, 0.01 μg/ml of adrenaline did not affect LPS-induced IL-6, IL-8, or PGE₂ production, but they were significantly increased at concentrations of 0.1 and 1 μg/ml. In the absence of LPS, H-89, dbcAMP, aminophylline, and adrenaline had no relevance to IL-6, IL-8, or PGE₂ production. These results suggest that the PKA pathway, and also PKA-activating drugs, enhance LPS-induced IL-6, IL-8, and PGE₂ production by HGFs. However, aminophylline may not have an effect on the production of these molecules at concentrations used in clinical settings (8 to 20 μg/ml in serum). These results suggest that aminophylline does not affect inflammatory responses in periodontal disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
H-89 dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Aminophyllin, ≥98%, powder