Merck
  • The transcription factor HNF-4α: a key factor of the intestinal uptake of fatty acids in mouse.
The transcription factor HNF-4α: a key factor of the intestinal uptake of fatty acids in mouse.
American journal of physiology. Gastrointestinal and liver physiology (2012-03-31)
Vincent Frochot, Malik Alqub, Anne-Laure Cattin, Véronique Carrière, Anne Houllier, Floriane Baraille, Laurence Barbot, Susan Saint-Just, Agnès Ribeiro, Michel Lacasa, Philippe Cardot, Jean Chambaz, Monique Rousset, Jean-Marc Lacorte
ZUSAMMENFASSUNG
With an excessive postprandial accumulation of intestine-derived, triglyceride-rich lipoproteins being a risk factor of cardiovascular diseases, it is essential to characterize the mechanisms controlling the intestinal absorption of dietary lipids. Our aim was to investigate the role of the transcription factor hepatocyte nuclear factor (HNF)-4α in this process. We used transgenic mice with a specific and inducible intestinal knockout of Hnf-4α gene. One hour after a lipid bolus, in the presence of the lipase inhibitor tyloxapol, lower amounts of triglycerides were found in both plasma and intestinal epithelium of the intestine-specific Hnf-4α knockout (Hnf-4α(intΔ)) mice compared with the Hnf-4α(loxP/loxP) control mice. These discrepancies were due to a net decrease of the intestinal uptake of fatty acid in Hnf-4α(intΔ) mice compared with Hnf-4α(loxP/loxP) mice, as assessed by the amount of radioactivity that was recovered in intestine and plasma after gavage with labeled triolein or oleic acid, or in intestinal epithelial cells isolated from jejunum after a supply of labeled oleic acid-containing micelles. This decreased fatty acid uptake was associated with significant lower levels of the fatty acid transport protein-4 mRNA and protein along the intestinal tract and with a lower acyl-CoA synthetase activity in Hnf-4α(intΔ) mice compared with the control mice. We conclude that the transcription factor HNF-4α is a key factor of the intestinal absorption of dietary lipids, which controls this process as early as in the initial step of fatty acid uptake by enterocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyloxapol, nonionic surfactant
Sigma-Aldrich
Tyloxapol, BioXtra
Sigma-Aldrich
MTP Activity Assay Kit, Supplied by Roar Biomedical, Inc.