Merck
  • Intravascular infusion of lidocaine: a novel way to relieve sudden internal carotid artery occlusion in embolization of intracranial aneurysms.
Intravascular infusion of lidocaine: a novel way to relieve sudden internal carotid artery occlusion in embolization of intracranial aneurysms.
Journal of neurological surgery. Part A, Central European neurosurgery (2012-04-03)
Qiao Yu Li, Wen Lin Xu, Yan Zhang, Pej Shan Lu, Zhi Cheng Yuan, Li Ping Zhan, Peng Wang, Xia Yu Lu, Bo Cheng
ZUSAMMENFASSUNG
Sudden internal carotid artery (ICA) occlusive vasospasm is a serious complication of intracranial aneurysm embolization. Conventional spasmolysis with papaverine yields a generally poor outcome. We believe that arterial infusion of lidocaine may offer a better outcome. We retrospectively reviewed the outcome of patients treated with either papaverine or lidocaine infusion for vasospasm during embolization. 14 patients undergoing intracranial aneurysm embolization had a ICA occlusive vasospasm. Among the 8 patients who received conventional treatment with papaverine the vasospasm improved partially in 5. In 3 cases, treatment was ineffective. 6 of the patients died within 3 days. 2 patients developed hemispheric infarction and underwent a decompressive craniectomy and subtotal resection of the infarct; 1 of these 2 patients died after 4 months and the other was severely disabled. In the 6 patients treated with lidocaine, spasmolysis and subsequent aneurysm treatment was successful in 5. In 1 patient who had preoperative stenosis of the carotid artery proximal to the aneurysm spasmolysis failed. ICA occlusive spasm is an extremely serious and often lethal complication in embolization of intracranial aneurysms. Conventional treatment with papaverine has a poor outcome, whereas arterial infusion of lidocaine may achieve better results.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Papaverin -hydrochlorid, powder
Supelco
Papaverin -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Papaverin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard