Merck
Tea tree oil as a novel antipsoriasis weapon.
Skin pharmacology and physiology (2012-04-05)
Nader Pazyar, Reza Yaghoobi
ZUSAMMENFASSUNG
Psoriasis is a clinical skin disease that is characterized by erythematous scaling plaques and involves the extensor site of the extremities, the scalp and other surfaces of the skin. Tea tree oil (TTO) is considered an essential oil, obtained by steam distillation of the leaves and terminal branchlets of Melaleuca alternifolia. Notably,terpinen-4-ol, the major TTO constituent, has been found to have potent anti-inflammatory properties. It is suggested that terpinen-4-ol may be a novel potential agent against psoriasis. This article draws attention to the antipsoriatic effect of TTO and provides a theoretical molecular approach.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Teebaumöl, FG