Merck
  • Identification of trace organic pollutants in drinking water and the associated human health risks in Jiangsu Province, China.
Identification of trace organic pollutants in drinking water and the associated human health risks in Jiangsu Province, China.
Bulletin of environmental contamination and toxicology (2012-04-06)
Bing Li, Changsheng Qu, Jun Bi
ZUSAMMENFASSUNG
Organochlorine pesticides (OCPs), polycyclic aromatic hydrocarbons (PAHs), dibutyl phthalate (DNBP) and di-2-ethylhexyl phthalate (DEHP) were all detectable in surface water in Jiangsu Province, China. Concentrations of OCPs ranged from 5.13 to 8.15 ng/L. PAHs were found ranging from 14.7 to 24.5 ng/L. Concentrations of DNBP and DEHP ranged from 16 to 5,857.5 ng/L and 556 to 15,670.7 ng/L, respectively. Greater than 70 % of chemicals were removed in water treatment processes. The carcinogenic risks posed by trace organic pollutants through tap water ingestion were lower than 10(-6), and the noncarcinogenic risks were less than 10(-5).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard
Supelco
2,4-Dinitrophenol -Lösung, reference material, 5000 μg/mL in methanol
Supelco
Dinoseb, PESTANAL®, analytical standard