Merck
  • Molecular recognition of T:G mismatched base pairs in DNA as studied by electrospray ionization mass spectrometry.
Molecular recognition of T:G mismatched base pairs in DNA as studied by electrospray ionization mass spectrometry.
ChemMedChem (2012-04-11)
Federico Riccardi Sirtori, Giancarlo Aldini, Maristella Colombo, Nicoletta Colombo, Jan Malyszko, Giulio Vistoli, Roberto D'Alessio
ZUSAMMENFASSUNG
Postreplicative mismatch repair (MMR) is a cellular system involved in the recognition and correction of DNA polymerase errors that escape detection in proofreading. Of the various mismatched bases, T:G pairing in DNA is one of the more common mutations leading to the formation of tumors in humans. In addition, the absence of the MMR system can generate resistance to several chemotherapeutic agents, particularly DNA-damaging substances. The main purpose of this study was the setup and validation of an electrospray ionization (ESI) mass spectrometry method for the identification of small molecules that are able to recognize T:G mismatches in DNA targets. These findings could be useful for the discovery of new antitumor drugs. The analytical method is based on the ability of electrospray to preserve the noncovalent adducts present in solution and transfer them to the gas phase. Lexitropsin derivatives (polyimidazole compounds) have been previously described as selective for T:G mismatch binding by NMR and ITC studies. We synthesized and tested various polyimidazole derivatives, one of which in particular (NMS-057) showed a higher affinity for an oligonucleotide DNA sequence containing a T:G mismatched base pair. To rationalize these findings, molecular docking studies were performed using available NMR structures. Moreover, ESI-MS experiments, performed on an orbitrap mass spectrometer, highlighted the formation of heterodimeric complexes between DNA sequences, distamycin A, and polyimidazole compounds. Our results confirm that this ESI method could be a valuable tool for the identification of new molecules able to specifically recognize T:G mismatched base pairs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Netropsin -dihydrochlorid, from Streptomyces netropsis, ≥98% (HPLC), powder