Merck
Evaluating a novel analgesic strategy for ring castration of ram lambs.
Veterinary anaesthesia and analgesia (2012-04-25)
David R Paull, Alison H Small, Caroline Lee, Pierre Palladin, Ian G Colditz
ZUSAMMENFASSUNG
To evaluate the analgesic efficacy of the NSAIDs flunixin and meloxicam administered locally to the scrotum before ring castration. Randomised, controlled, prospective study. Forty eight single born male Merino lambs. Lambs, aged approximately 4 weeks, were allocated to four groups for castration. Groups were: sham control; castration + saline; castration + flunixin; castration + meloxicam. Drugs (5 mL) were administered subcutaneously around the circumference of the scrotum immediately before castration. Cortisol, rectal temperature, haematology and plasma haptoglobin were measured before and up to 48 hours after treatment. Behaviour recorded by video for 12 hours after treatment was classified as pain avoidance behaviours in the first hour and postural behaviours in three 4 hour intervals. Ring castration (saline group) induced a bi-phasic increase in cortisol with peaks at 90 minutes and 24 hours but no significant changes in haematology, haptoglobin or rectal temperature. Pain avoidance behaviours were increased and teat seeking decreased. Normal lying and normal standing postures were decreased and abnormal ventral lying, statue standing, abnormal standing and total abnormal postures increased. Flunixin decreased cortisol at 90 minutes (60.3 versus 117.3 nmol L(-1) ) and cortisol AUC (0-6 hours), decreased elevated leg movement (2.5 versus 5.4 events) and sum of pain avoidance behaviours (8.5 versus 16.7 events), improved time spent in normal ventral lying and decreased abnormal ventral lying and total abnormal postures compared to saline treated lambs. In a similar contrast, meloxicam caused non-significant decreases in cortisol at 90 minutes, cortisol AUC (0-6 hours) and pain avoidance behaviours, and significantly improved the postural behaviours normal ventral lying (26.7 versus 15.4%) and normal standing (13.9 versus 7.5%), and reduced abnormal standing and total abnormal postures. Physiological and behavioural responses associated with ring castration for both NSAID treatment groups were generally greater than sham controls. Locally administered NSAIDs provided partial analgesia for ring castration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Flunixin, VETRANAL®, analytical standard
Sigma-Aldrich
Clonixin, ≥98% (HPLC)