Merck
  • [Effects of advanced glycosylation end products and tetrandrine on proliferation of K562 and K562/A02 cells].
[Effects of advanced glycosylation end products and tetrandrine on proliferation of K562 and K562/A02 cells].
Zhongguo shi yan xue ye xue za zhi (2012-05-01)
Tian-Tian Wang, Dan Yan, Bao-An Chen, Jian Wang, Guo-Hua Xia, Shuai Wang, Jian Cheng, Jia-Hua Ding, Wen Bao
ZUSAMMENFASSUNG
This study was aimed to investigate the effect of advanced glycosylation end products (AGE) on the proliferation of K562 and K562/A02 cells, the effect of tetrandrine (Tet) on proliferation of K562 and K562/A02 cells induced by AGE, and their mechanisms. The effects of AGE on proliferation of K562 and K562/A02 cells and Tet on the proliferation of AGE-induced K562 and K562/A02 cells were assayed by CCK8 kit, the apoptosis rate and the expression of receptor of advanced glycosylation end products (RAGE) in K562 and K562/A02 cells were determined by flow cytometry, the expression of RAGE mRNA was detected by semi-quantitative RT-PCR. The results showed that AGE could promote the proliferation of K562 and K562/A02 cells in a concentration-dependent manner, the cell proliferation was enhanced with time increasing in 0 - 48 h, and was higher than control group after 72 h. AGE up-regulated the RAGE mRNA and protein expressions of K562 and K562/A02 cells in a concentration-dependent manner. Treatment of Tet combined with AGE for 48 h could inhibit the proliferation of K562 and K562/A02 cells promoted by AGE in a concentration-dependent manner, which probably by inducing cell apoptosis, however, there was no obvious effect in the up-regulating expression of RAGE mRNA and protein induced by AGE. It is concluded that AGE can promote the proliferation of K562 and K562/A02 cells, which is probably induced by up-regulating the expression of RAGE mRNA and protein. Tet can inhibit the proliferation of K562 and K562/A02 cells induced by AGE, and the mechanism may be not closely associated with changes of the up-regulating expression of RAGE mRNA and protein induced by AGE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Tetrandrin, analytical standard, for drug analysis