Merck
Efficient activation of thioglycosides with N-(p-methylphenylthio)-ε-caprolactam-TMSOTf.
Carbohydrate research (2012-05-01)
Sajal Kumar Maity, Nabamita Basu, Rina Ghosh
ZUSAMMENFASSUNG
N-(p-Methylphenylthio)-ε-caprolactam (1) in combination with trimethylsilyl trifluoromethanesulfonate (TMSOTf) provides an efficient thiophilic promoter system, capable of activating different thioglycosides. Both 'armed' and 'disarmed' thioglycosyl donors were activated for glycosidic bond formation. Notably, this reagent combination works well in reactivity-based one-pot oligosaccharide assembly strategy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trimethylsilyltrifluormethansulfonat, 99%
Sigma-Aldrich
Trimethylsilyltrifluormethansulfonat, purum, ≥98.0% (T)