Merck
  • Mixed metal copper(II)-nickel(II) and copper(II)-zinc(II) complexes of multihistidine peptide fragments of human prion protein.
Mixed metal copper(II)-nickel(II) and copper(II)-zinc(II) complexes of multihistidine peptide fragments of human prion protein.
Journal of inorganic biochemistry (2012-05-01)
Viktória Jószai, Ildikó Turi, Csilla Kállay, Giuseppe Pappalardo, Giuseppe Di Natale, Enrico Rizzarelli, Imre Sóvágó
ZUSAMMENFASSUNG
Mixed metal copper(II)-nickel(II) and copper(II)-zinc(II) complexes of four peptide fragments of human prion protein have been studied by potentiometric, UV-vis and circular dichroism spectroscopic techniques. One peptide contained three histidyl residues: HuPrP(84-114) with H85 inside and H96, H111 outside the octarepeat domain. The other three peptides contained two histidyl residues; H96 and H111 for HuPrP(91-115) and HuPrP(84-114)H85A while HuPrP(84-114)H96A contained the histidyl residues at positions 85 and 111. It was found that both histidines of the latter peptides can simultaneously bind copper(II) and nickel(II) ions and dinuclear mixed metal complexes can exist in slightly alkaline solution. One molecule of the peptide with three histidyl residues can bind two copper(II) and one nickel(II) ions. H85 and H111 were identified as the major copper(II) and H96 as the preferred nickel(II) binding sites in mixed metal species. The studies on the zinc(II)-PrP peptide binary systems revealed that zinc(II) ions can coordinate to the 31-mer PrP peptide fragments in the form of macrochelates with two or three coordinated imidazol-nitrogens but the low stability of these complexes cannot prevent the hydrolysis of the metal ion in slightly alkaline solution. These data provide further support for the outstanding affinity of copper(II) ions towards the peptide fragments of prion protein but the binding of nickel(II) can significantly modify the distribution of copper(II) among the available metal binding sites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, ReagentPlus®
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Nickel(II)-chlorid, 98%
Sigma-Aldrich
Kupfer(II)-chlorid, 97%
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, reagent grade
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, 99.9% trace metals basis
Sigma-Aldrich
Kupfer(II)-chlorid, powder, 99%
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, puriss. p.a., ≥98%
Sigma-Aldrich
Kupfer(II)-chlorid, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, BioReagent, suitable for cell culture
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nickel(II)-chlorid, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Kupfer(II)-chlorid, 99.999% trace metals basis
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, 99.999%
Sigma-Aldrich
Nickel(II)-chlorid Hydrat, 99.95% trace metals basis
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, 99.999% trace metals basis
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, ≥99.95% trace metals basis