Merck
Crystal structure of dihydroorotate dehydrogenase from Leishmania major.
Biochimie (2012-05-01)
Artur T Cordeiro, Patricia R Feliciano, Matheus P Pinheiro, M Cristina Nonato
ZUSAMMENFASSUNG
Dihydroorotate dehydrogenase (DHODH) is the fourth enzyme in the de novo pyrimidine biosynthetic pathway and has been exploited as the target for therapy against proliferative and parasitic diseases. In this study, we report the crystal structures of DHODH from Leishmania major, the species of Leishmania associated with zoonotic cutaneous leishmaniasis, in its apo form and in complex with orotate and fumarate molecules. Both orotate and fumarate were found to bind to the same active site and exploit similar interactions, consistent with a ping-pong mechanism described for class 1A DHODHs. Analysis of LmDHODH structures reveals that rearrangements in the conformation of the catalytic loop have direct influence on the dimeric interface. This is the first structural evidence of a relationship between the dimeric form and the catalytic mechanism. According to our analysis, the high sequence and structural similarity observed among trypanosomatid DHODH suggest that a single strategy of structure-based inhibitor design can be used to validate DHODH as a druggable target against multiple neglected tropical diseases such as Leishmaniasis, Sleeping sickness and Chagas' diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fumarsäure, ≥99.0% (T)
Sigma-Aldrich
Orotsäure, ≥98% (titration), anhydrous
Sigma-Aldrich
Fumarsäure, FCC, FG
Fumarsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Fumarsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland