Merck
  • Supported Au nanoparticles as efficient catalysts for aerobic homocoupling of phenylboronic acid.
Supported Au nanoparticles as efficient catalysts for aerobic homocoupling of phenylboronic acid.
Chemical communications (Cambridge, England) (2012-05-01)
Liang Wang, Wei Zhang, Dang Sheng Su, Xiangju Meng, Feng-Shou Xiao
ZUSAMMENFASSUNG
Au nanoparticles with small sizes (1-4 nm) were effectively formed on Mg-Al mixed oxides (Au/MAO), which showed superior catalytic performances and good recyclability in aerobic homocoupling of phenylboronic acid.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenylboronsäure, 95%
Sigma-Aldrich
Phenylboronsäure, purum, ≥97.0% (HPLC)