Merck
  • Efficient photocatalytic hydrogen production by platinum-loaded carbon-doped cadmium indate nanoparticles.
Efficient photocatalytic hydrogen production by platinum-loaded carbon-doped cadmium indate nanoparticles.
ACS applied materials & interfaces (2012-05-04)
Jason M Thornton, Daniel Raftery
ZUSAMMENFASSUNG
Undoped and carbon doped cadmium indate (CdIn(2)O(4)) powders were synthesized using a sol-gel pyrolysis method and evaluated for hydrogen generation activity under UV-visible irradiation without the use of a sacrificial reagent. Each catalyst powder was loaded with a platinum cocatalyst in order to increase electron-hole pair separation and promote surface reactions. Carbon-doped indium oxide and cadmium oxide were also prepared and analyzed for comparison. UV-vis diffuse reflectance spectra indicate the band gap for C-CdIn(2)O(4) to be 2.3 eV. C-doped In(2)O(4) showed a hydrogen generation rate approximately double that of the undoped material. When compared to platinized TiO(2) in methanol, which was used as a control material, C-CdIn(2)O(4) showed a 4-fold increase in hydrogen production. The quantum efficiency of the material was calculated at different wavelength intervals and found to be 8.7% at 420-440 nm. The material was capable of hydrogen generation using visible light only and with good efficiency even at 510 nm.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cadmiumoxid, ≥99.99% trace metals basis
Sigma-Aldrich
Indium(III)-oxid, 99.99% trace metals basis
Sigma-Aldrich
Cadmiumoxid, powder, 99.5% trace metals basis
Sigma-Aldrich
Indium(III)-oxid, 99.998% trace metals basis
Sigma-Aldrich
Indium(III)-oxid, nanopowder, <100 nm particle size (TEM), 99.9% trace metals basis