Merck
Etiology of inflammatory bowel disease: a unified hypothesis.
World journal of gastroenterology (2012-05-04)
Xiaofa Qin
ZUSAMMENFASSUNG
Inflammatory bowel disease (IBD), including both ulcerative colitis (UC) and Crohn's disease (CD), emerged and dramatically increased for about a century. Despite extensive research, its cause remains regarded as unknown. About a decade ago, a series of findings made me suspect that saccharin may be a key causative factor for IBD, through its inhibition on gut bacteria and the resultant impaired inactivation of digestive proteases and over digestion of the mucus layer and gut barrier (the Bacteria-Protease-Mucus-Barrier hypothesis). It explained many puzzles in IBD such as its emergence and temporal changes in last century. Recently I further found evidence suggesting sucralose may be also linked to IBD through a similar mechanism as saccharin and have contributed to the recent worldwide increase of IBD. This new hypothesis suggests that UC and CD are just two symptoms of the same morbidity, rather than two different diseases. They are both caused by a weakening in gut barrier and only differ in that UC is mainly due to increased infiltration of gut bacteria and the resultant recruitment of neutrophils and formation of crypt abscess, while CD is mainly due to increased infiltration of antigens and particles from gut lumen and the resultant recruitment of macrophages and formation of granulomas. It explained the delayed appearance but accelerated increase of CD over UC and many other phenomena. This paper aims to provide a detailed description of a unified hypothesis regarding the etiology of IBD, including the cause and mechanism of IBD, as well as the relationship between UC and CD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Supelco
Sucralose, analytical standard