Merck
  • Synthesis, molecular properties prediction, and anti-staphylococcal activity of N-acylhydrazones and new 1,3,4-oxadiazole derivatives.
Synthesis, molecular properties prediction, and anti-staphylococcal activity of N-acylhydrazones and new 1,3,4-oxadiazole derivatives.
Molecules (Basel, Switzerland) (2012-05-05)
Cledualdo Soares de Oliveira, Bruno Freitas Lira, Vivyanne dos Santos Falcão-Silva, Jose Pinto Siqueira, Jose Maria Barbosa-Filho, Petronio Filgueiras de Athayde-Filho
ZUSAMMENFASSUNG
Five new 1-(2-(5-nitrofuran-2-yl)-5-(aryl)-1,3,4-oxadiazol-3-(2H)-yl) ethanone compounds 5a-e were synthesized by cyclization of N-acylhydrazones 4a-e with acetic anhydride under reflux conditions. Their structures were fully characterized by IR, ¹H-NMR, and ¹³C-NMR. Furthermore, evaluations of the antibacterial activity of the 1,3,4-oxadiazoles 5a-e and N-acylhydrazones 4a-e showed strong activity against several strains of Staphylococcus aureus, with MICs between 4 μg/mL to 32 μg/mL. In silico studies of the parameters of Lipinski's Rule of Five, as well as the topological polar surface area (TPSA), absorption percentage (% ABS), drug likeness and drug score indicate that these compounds, especially 4a and 5d, have potential to be new drug candidates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäureanhydrid, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäureanhydrid, ACS reagent, ≥98.0%
Sigma-Aldrich
Essigsäureanhydrid, 99.5%
Supelco
Essigsäureanhydrid, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
Essigsäureanhydrid, Arxada quality, ≥99.5% (GC)