Merck
  • On-farm bioremediation of dimethazone and trifluralin residues in runoff water from an agricultural field.
On-farm bioremediation of dimethazone and trifluralin residues in runoff water from an agricultural field.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2012-05-09)
George F Antonious
ZUSAMMENFASSUNG
Bioremediation is the use of living organisms, primarily microorganisms, to degrade environmental contaminants into less toxic forms. Nine biobeds (ground cavity filled with a mixture of composted organic matter, topsoil, and a surface grass) were established at Kentucky State University research farm (Franklin County, KY) to study the impact of this practice on reducing surface runoff water contamination by residues of dimethazone and trifluralin herbicides arising from an agricultural field. Biobed (biofilter) systems were installed at the bottom of the slope of specially designed runoff plots to examine herbicides retention and degradation before entering streams and rivers. In addition to biobed systems, three soil management practices: municipal sewage sludge (SS), SS mixed with yard waste compost (SS + YW), and no-mulch rototilled bare soil (NM used for comparison purposes) were used to monitor the impact of soil amendments on herbicide residues in soil following natural rainfall events. Organic amendments increased soil organic matter content and herbicide residues retained in soil following rainfall events. Biobeds installed in NM soil reduced dimethazone and trifluralin by 84 and 82%, respectively in runoff water that would have been transported down the land slope of agricultural fields and contaminated natural water resources. Biobeds installed in SS and SS+YW treatments reduced dimethazone by 65 and 46% and trifluralin by 52 and 79%, respectively. These findings indicated that biobeds are effective for treating dimethazone and trifluralin residues in runoff water.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Clomazon, PESTANAL®, analytical standard
Supelco
Trifluralin, PESTANAL®, analytical standard