Merck
  • Plant bio-transformable HMG-CoA reductase gene loaded calcium phosphate nanoparticle: in vitro characterization and stability study.
Plant bio-transformable HMG-CoA reductase gene loaded calcium phosphate nanoparticle: in vitro characterization and stability study.
Current drug discovery technologies (2012-05-09)
Mehrnaz S Ohadi R, Amene Alvari, M Samim, Malik Z Abdin
ZUSAMMENFASSUNG
Encapsulation of plasmid DNA in nanoparticle is expected to enhance the stability of DNA, reproducibility and frequency of the genetic transformation in plants. Here we report the formulation of HMG Co-A reductase gene loaded calcium phosphate nanoparticles (Cap nanoparticles) and their in-vitro, in-vivo characterization. The developed Cap nanoparticles were characterized by DSC, FT-IR, and XRD. Developed Cap nanoparticles were spherical in shape having the particle size and zeta potential in the range of 10.86±0.09nm to 33.42±0.18nm and -25.5±0.07mV to -31.7±0.07mV (for Cap-I to Cap-IV). DNA releasing in acidic media showed, initially slow release followed by fast release with a maximum release of Cap-I (95.77±1.39%) > Cap-II (87.32±2.07%) > Cap-III (76.54±2.01%) > Cap-IV (72.93±1.75%) over 60min. Cap nanoparticles were quite stable at storage condition of 40±0.5°C/75±5%RH, 25±0.5°C/60±RH, 4±0.5°C/ambient humidity and the integrity of pDNA encapsulated was confirmed by gel electrophoresis. Compared to wild type C. intybus, transformation efficiency and enhanced biosynthesis of esculin with the DNA nanoparticles in C. intybus were about 10% and 71%, respectively. Antioxidant activity capacity of the biotransformed plants was significantly higher than the normal plant due to high accumulation of esculin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Esculin Hydrat, ≥98%
Sigma-Aldrich
Esculin Hydrat, ≥98.0% (HPLC)