Merck
Enantioselectivity in degradation and transformation of quizalofop-ethyl in soils.
Chirality (2012-05-09)
Zhaoyang Li, Qiaoling Li, Fengning Cheng, Wenshu Zhang, Weixiao Wang, Jingyin Li
ZUSAMMENFASSUNG
The enantioselective degradation of quizalofop-ethyl and its metabolite quizalofop-acid in two soils, a Wuhan acidic soil and a Baoding alkaline soil, was investigated. The dissipation of quizalofop-ethyl consisted of two phases, a rapidly deceasing first phase that lasted 1 day and a slowly decreasing second phase that extended till the end of the incubation. It is shown that S-quizalofop-ethyl degraded slightly faster than R-quizalofop-ethyl in the two soils. Further incubation of enantiopure enaniomers showed that quizalofop-ethyl was configurationally stable in soil. Quizalofop-acid was produced quickly, and its amount reached a maximum at 1-6 days time and then decreased slowly with half-lives ranging from 11 to 21 days. The results also showed that quizalofop-acid degraded faster in the acidic Wuhan soil than in the alkaline Baoding soil. At last, significant enantiomerization from S-quizalofop-acid to R-quizalofop-acid was observed, and the enantiomerization was fast, resulting in residues enriched with R-quizalofop-acid whatever racemic quizalofop-ethyl or pure enantiomers were initially applied in the soils.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Quizalofop-P-ethyl, PESTANAL®, analytical standard
Supelco
Quizalofopethyl, PESTANAL®, analytical standard