- Gas-chromatographic method of analysis for urinary organic acids. I. Retention indices of 155 metabolically important compounds.
Gas-chromatographic method of analysis for urinary organic acids. I. Retention indices of 155 metabolically important compounds.
Clinical chemistry (1980-12-01)
K Tanaka, D G Hine, A West-Dull, T B Lynn
PMID7438429
ZUSAMMENFASSUNG
Gas-chromatographic retention indices are given, in terms of methylene units, for 155 metabolically important compounds (mostly organic acids) as trimethylsilyl derivatives on 10% OV-1 and 10% OV-17 columns. Comprehensive references on metabolic diseases that can be diagnosed by detection of these metabolites are cross-indexed to facilitate the use of the methylene-unit list. With the data presented here, it is now possible to diagnose more than 25 well-defined organic acidurias by use of gas chromatography alone.