Merck
Organ accumulation in mice after inhalation of single or mixed essential oil compounds.
Phytotherapy research : PTR (2012-05-15)
Tadaaki Satou, Mizuho Takahashi, Hikaru Kasuya, Shio Murakami, Shinichiro Hayashi, Kiyomi Sadamoto, Kazuo Koike
ZUSAMMENFASSUNG
Essential oils are composed of multiple components. It is thought that the effect of essential oils is due to specific component ratios, which may differ from the original ratio when the essential oil is absorbed. However, very little detailed research exists in this area. We studied the distribution of essential oil components after inhalation of single and mixed components in mice. This research was done using four main components of Alpinia zerumbet (Pers.) B. L. Burtt. and R. M. Sm.: α-pinene, p-cymene, 1,8-cineole, and limonene. After inhalation of single or mixed components for 90 min, component levels in the brain and liver of mice were measured. The results indicated that the amount of α-pinene in the brain and liver was twofold greater after mixed-component inhalation than that after single-component inhalation. In a comparison of the components of the mixed inhalation, the ratio of α-pinene increased to about three times that of 1,8-cineole. It is thought that the absorption via the nasal mucus greatly influences this phenomenon. The results of this investigation of the bodily distribution of essential oil volatile components may provide clues for elucidating their action.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eucalyptol, 99%
Sigma-Aldrich
α-Pinen, 98%
Sigma-Aldrich
p-Cymol, 99%
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Supelco
Eucalyptol, analytical standard
Sigma-Aldrich
p-Cymol, ≥97%, FG
Supelco
1,8-Cineol, primary reference standard
Supelco
p-Cymol, analytical standard
α-Pinen, primary reference standard
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.
Cineol, European Pharmacopoeia (EP) Reference Standard