Merck
  • Effect of dietary supplements in reducing probability of death for uremic crises in dogs affected by chronic kidney disease (masked RCCT).
Effect of dietary supplements in reducing probability of death for uremic crises in dogs affected by chronic kidney disease (masked RCCT).
TheScientificWorldJournal (2012-05-18)
Andrea Zatelli, Marco Pierantozzi, Paola D'Ippolito, Mauro Bigliati, Eric Zini
ZUSAMMENFASSUNG
Chitosan and alkalinizing agents can decrease morbidity and mortality in humans with chronic kidney disease (CKD). Whether this holds true in dog is not known. Objective of the study was to determine whether a commercial dietary supplement containing chitosan, phosphate binders, and alkalinizing agents (Renal), compared to placebo, reduces mortality rate due to uremic crises in dogs with spontaneous CKD, fed a renal diet (RD). A masked RCCT was performed including 31 azotemic dogs with spontaneous CKD. Dogs enrolled in the study were randomly allocated to receive RD plus placebo (group A; 15 dogs) or RD plus Renal (group B; 16 dogs). During a first 4-week period, all dogs were fed an RD and then randomized and clinically evaluated up to 44 weeks. The effects of dietary supplements on mortality rate due to uremic crises were assessed. At 44 weeks, compared to group A, dogs in group B had approximately 50% lower mortality rate due to uremic crises (P = 0.015). Dietary supplementation with chitosan, phosphate binders, and alkalinizing agents, along with an RD, is beneficial in reducing mortality rate in dogs with spontaneous CKD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumcitrat tribasisch Monohydrat, ≥98% (GC/titration)
Sigma-Aldrich
Kaliumcitrat Monohydrat, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Kaliumcitrat Monohydrat, meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)
Sigma-Aldrich
Kaliumcitrat Monohydrat, meets USP testing specifications
Sigma-Aldrich
Kaliumcitrat tribasisch Monohydrat, suitable for cell culture