Merck
  • Efficacy of pentane, toluene, and benzene to support aerobic cometabolism of ethylene dibromide.
Efficacy of pentane, toluene, and benzene to support aerobic cometabolism of ethylene dibromide.
New biotechnology (2012-05-23)
Anthony S Danko, Patrícia O Leitão, Matthew F Verce, David L Freedman
ZUSAMMENFASSUNG
The ability of pentane, benzene, and toluene to support aerobic cometabolism of ethylene dibromide (1,2-dibromoethane, EDB) was evaluated. A pentane enrichment culture cometabolized EDB, with a transformation capacity of 0.35 μmol EDB/mg biomass (66.2 μg EDB/mg biomass) in the absence of growth substrate. It also cometabolized EDB while actively growing on pentane. However, enrichment cultures grown on benzene or toluene could not cometabolize EDB, with or without their respective growth substrates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Sigma-Aldrich
Pentan, ≥99% (GC)
Sigma-Aldrich
Pentan, anhydrous, ≥99%
Sigma-Aldrich
1,2-Dibromethan, 98%
Sigma-Aldrich
1,2-Dibromethan, ≥99%
Supelco
1,2-Dichlorethan, analytical standard
Supelco
1,2–Dichlorethan -Lösung, certified reference material, 200 μg/mL in methanol
Sigma-Aldrich
1,2-Dibromethan, purum, ≥98.0% (GC)
Supelco
Pentan, analytical standard
Supelco
1,2-Dibromethan -Lösung, certified reference material, 200 μg/mL in methanol