- A ligand-free Pd-catalyzed cascade reaction: an access to the highly diverse isoquinolin-1(2H)-one derivatives via isocyanide and Ugi-MCR synthesized amide precursors.
A ligand-free Pd-catalyzed cascade reaction: an access to the highly diverse isoquinolin-1(2H)-one derivatives via isocyanide and Ugi-MCR synthesized amide precursors.
Organic letters (2012-05-26)
Vikas Tyagi, Shahnawaz Khan, Archana Giri, Harsh M Gauniyal, B Sridhar, Prem M S Chauhan
PMID22625424
ZUSAMMENFASSUNG
A novel ligand-free palladium-catalyzed cascade reaction for the synthesis of highly diverse isoquinolin-1(2H)-one derivatives from isocyanide and amide precursors synthesized by Ugi-MCR has been developed. A broad variety of acids, amines, and isocyanides were used as starting materials for Ugi-MCR leading to various amide precursors, which in turn provided entry into diverse isoquinolin-1(2H)-one derivatives. The reaction proceeds through tandem isocyanide insertion with intramolecular cyclization followed by a Mazurciewitcz-Ganesan type sequence to provide isoquinoline-1(2H)-one derivatives in moderate to good yields.
MATERIALIEN