Merck
Management of cluster headache.
CNS drugs (2012-06-02)
Peer C Tfelt-Hansen, Rigmor H Jensen
ZUSAMMENFASSUNG
The prevalence of cluster headache is 0.1% and cluster headache is often not diagnosed or misdiagnosed as migraine or sinusitis. In cluster headache there is often a considerable diagnostic delay - an average of 7 years in a population-based survey. Cluster headache is characterized by very severe or severe orbital or periorbital pain with a duration of 15-180 minutes. The cluster headache attacks are accompanied by characteristic associated unilateral symptoms such as tearing, nasal congestion and/or rhinorrhoea, eyelid oedema, miosis and/or ptosis. In addition, there is a sense of restlessness and agitation. Patients may have up to eight attacks per day. Episodic cluster headache (ECH) occurs in clusters of weeks to months duration, whereas chronic cluster headache (CCH) attacks occur for more than 1 year without remissions. Management of cluster headache is divided into acute attack treatment and prophylactic treatment. In ECH and CCH the attacks can be treated with oxygen (12 L/min) or subcutaneous sumatriptan 6 mg. For both oxygen and sumatriptan there are two randomized, placebo-controlled trials demonstrating efficacy. In both ECH and CCH, verapamil is the prophylactic drug of choice. Verapamil 360 mg/day was found to be superior to placebo in one clinical trial. In clinical practice, daily doses of 480-720 mg are mostly used. Thus, the dose of verapamil used in cluster headache treatment may be double the dose used in cardiology, and with the higher doses the PR interval should be checked with an ECG. At the start of a cluster, transitional preventive treatment such as corticosteroids or greater occipital nerve blockade can be given. In CCH and in long-standing clusters of ECH, lithium, methysergide, topiramate, valproic acid and ergotamine tartrate can be used as add-on prophylactic treatment. In drug-resistant CCH, neuromodulation with either occipital nerve stimulation or deep brain stimulation of the hypothalamus is an alternative treatment strategy. For most cluster headache patients there are fairly good treatment options both for acute attacks and for prophylaxis. The big problem is the diagnosis of cluster headache as demonstrated by the diagnostic delay of 7 years. However, the relatively short-lasting attack of pain in one eye with typical associated symptoms should lead the family doctor to suspect cluster headache resulting in a referral to a neurologist or a headache centre with experience in the treatment of cluster headache.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ergotamin -D-tartrat, ≥97.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Methysergid -maleat (Salz), solid