Merck
  • Signal amplification architecture for electrochemical aptasensor based on network-like thiocyanuric acid/gold nanoparticle/ssDNA.
Signal amplification architecture for electrochemical aptasensor based on network-like thiocyanuric acid/gold nanoparticle/ssDNA.
Biosensors & bioelectronics (2012-06-02)
Zhengbo Chen, Lidong Li, Yu Tian, Xiaojiao Mu, Lin Guo
ZUSAMMENFASSUNG
In this work, we described signal amplification architecture for electronic aptamer-based sensor (E-AB), which is applicable to a wide range of aptamers. Herein, we only take lysozyme as the representative sensing target. The amplification method was based on the network of thiocyanuric acid (TCA)/gold nanoparticles (AuNPs) modified with ssDNA. The binding event can be detected by a decrease in the integrated charge of the surface-bound [Ru(NH(3))(6)](3+) which electrostatically absorbed onto the negatively charged phosphate backbones of DNA. In the presence of target molecules, a large amount of TCA/AuNP/ssDNA network associated with [Ru(NH(3))(6)](3+) would be removed from the electrode surface, leading to a significant decrease of redox current. Cyclic voltammetry (CV) signals of [Ru(NH(3))(6)](3+) provides quantitative measures of the concentrations of lysozyme, with a linear calibration ranging from 5 pM to 1 nM and a detection limit is 0.1 pM. The detection limit of the proposed sensor is one order of magnitude and three orders of magnitude more sensitive than the detection limits in the absence of TCA (5 pM) and in the absence of TCA/AuNP/ssDNA network (0.5 nM). This amplification method is promising for broad potential application in clinic assay and various protein analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyanursäure, 98%
Supelco
Cyanursäure, analytical standard