Merck
  • Effect of norspermidine and its related triamines on the cell-free polyphenylalanine synthesizing system from Vibrio parahaemolyticus.
Effect of norspermidine and its related triamines on the cell-free polyphenylalanine synthesizing system from Vibrio parahaemolyticus.
Microbiology and immunology (1990-01-01)
S Yamamoto, Y Koumoto, S Shikami, S Shinoda
ZUSAMMENFASSUNG
The effect of norspermidine and its structurally related triamines on the cell-free polyphenylalanine synthesizing system from Vibrio parahaemolyticus was examined in connection with the requirement of the system for monovalent cation. In the absence of norspermidine, the maximal incorporation of [14C]phenylalanine into hot trichloroacetic acid insoluble material was observed under ionic conditions of 12 mM Mg2+ and 50 mM NH4+. K+ could partially substitute for NH4+, but Na+ could not. The addition of norspermidine to the polyphenylalanine synthetic reaction mixture not only lowered the optimal Mg2+ concentration, but it also stimulated the polyphenylalanine synthesis up to 2-fold with no significant increase in misincorporation of [14C]leucine. Other triamines having one or two methylene chains more than norspermidine were also effective in eliciting these effects. Furthermore, Na+ could not support the polyphenylalanine synthesis even in the presence of norspermidine and, on the contrary, inhibited the polyphenylalanine synthesis induced by NH4+ regardless of whether norspermidine was present or not. These findings are discussed in comparison with the properties of other bacterial cell-free systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bis-(3-aminopropyl)-amin, 98%