Merck
  • The effect of strontium and combinations of strontium and fluoride on the remineralization of artificial caries lesions in vitro.
The effect of strontium and combinations of strontium and fluoride on the remineralization of artificial caries lesions in vitro.
Quintessence international (Berlin, Germany : 1985) (2012-06-07)
Ghaeth H Yassen, Frank Lippert, George Eckert, Jennifer Eder, Andrea Ferreira Zandoná
ZUSAMMENFASSUNG
To determine the effect of relatively low strontium concentrations on enamel remineralization and investigate the dose-response effects of strontium and fluoride combinations on the remineralization of artificial caries lesions in vitro. Artificial caries lesions were created in 135 bovine enamel specimens. Lesion severity was analyzed using transverse microradiography (TMR) and quantitative light-induced fluorescence (QLF). The specimens were randomly assigned to nine treatment groups based on lesion volume after lesion creation, as measured by TMR. Treatment groups were based on a 3 x 3 factorial design (0/0.05/0.1 ppm fluoride and 0/10/15 ppm strontium). Lesions were remineralized at 37°C for 14 days in artificial saliva, which was supplemented or not with NaF and/or SrCl2 x 6H2O. Lesion remineralization was assessed using QLF and TMR. Data were analyzed using ANOVA. For the TMR data, lesion remineralization in the 10 ppm strontium + 0.05 ppm fluoride group was significantly higher than in all other groups (P < .05) except the 0 ppm strontium + 0.05 ppm fluoride group (P = .06). The 10 ppm strontium + 0 ppm fluoride group exhibited significantly less remineralization than the 0 ppm strontium + 0 ppm fluoride group (P = .048). For the QLF data, intergroup differences were not the same as for the TMR analysis. The QLF measurement was only moderately correlated with TMR mineral loss (r = -0.37). Strontium alone did not improve the remineralization of artificial caries lesions under the chosen in vitro conditions. However, a synergistic effect between the combination of fluoride and strontium was found at specific concentrations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Strontiumchlorid Hexahydrat, ACS reagent, 99%
Sigma-Aldrich
Strontiumchlorid, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Strontiumchlorid Hexahydrat, 99.995% trace metals basis
Sigma-Aldrich
Strontiumchlorid -Lösung, 0.1 M