- Hydrolysis studies of phosphodichloridate and thiophosphodichloridate ions.
Hydrolysis studies of phosphodichloridate and thiophosphodichloridate ions.
The Journal of organic chemistry (2012-06-12)
Richard J Delley, AnnMarie C O'Donoghue, David R W Hodgson
PMID22681395
ZUSAMMENFASSUNG
We report the pH-k(obs) profiles for the hydrolyses of phosphodichloridate and thiophosphodichloridate ions in aqueous solutions. Both species show broad pH-k(obs) plateaus that extend to high pHs.