- Synthesis of tetrazole-fused glycosides by a tandem fragmentation-cyclization reaction.
Synthesis of tetrazole-fused glycosides by a tandem fragmentation-cyclization reaction.
Organic letters (2012-06-15)
Nieves R Paz, Andrés G Santana, Cosme G Francisco, Ernesto Suárez, Concepción C González
PMID22694091
ZUSAMMENFASSUNG
The fragmentation of anomeric alkoxyl radicals (ARF) and the subsequent intramolecular cyclization promoted by hypervalent iodine reagents provide an excellent method for the synthesis of tetrazolo-sugars. This new reaction offers additional advantages for the synthesis of these compounds, including the ready availability of the starting materials, experimental simplicity, mild conditions, and good yields.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetrazol -Lösung, suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile