Merck
  • Complexation of bovine serum albumin and sugar beet pectin: structural transitions and phase diagram.
Complexation of bovine serum albumin and sugar beet pectin: structural transitions and phase diagram.
Langmuir : the ACS journal of surfaces and colloids (2012-06-16)
Xiangyang Li, Yapeng Fang, Saphwan Al-Assaf, Glyn O Phillips, Xiaolin Yao, Yifeng Zhang, Meng Zhao, Ke Zhang, Fatang Jiang
ZUSAMMENFASSUNG
The complexation between bovine serum albumin (BSA) and sugar beet pectin (SBP) was studied in situ by coupling glucono-δ-lactone (GDL) induced acidification with dynamic light scattering and turbidity measurements. Individual measurements at specific pHs and mixing ratios were also carried out using zeta potentiometry, gel permeation chromatography-multiangle laser light scattering (GPC-MALLS), and isothermal titration calorimetry (ITC). These investigations together enabled the establishment of a phase diagram of BSA/SBP and the identification of the molecular events during protein/polysaccharide complexation in relation to the phase diagram, which showed five regions: (I) a stable region of mixed individual soluble polymers, (II) a stable region of intramolecular soluble complexes, (III) a quasi-stable region of intermolecular soluble complexes, (IV) an unstable region of intermolecular insoluble complexes, and (V) a second stable region of mixed individual soluble polymers, on lowering pH. We found for the first time that the complexation could take place well above the critical pH(c), the value that most previous studies had regarded as the onset occurrence of complexation. A model of structural transitions between the regions was proposed. The borderline between region II and region III represents the BSA/SBP stoichiometry for intramolecular soluble complex at a specific pH, while that between region III and region IV identifies the composition of the intermolecular insoluble complex. Also studied was the effect of NaCl and CaCl(2) on the phase diagram and structural transitions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Gluconsäure δ-Lacton, ≥99.0% (GC)
Sigma-Aldrich
Gluconolacton, 99.0-101.0%, meets USP testing specifications