Merck
Detection of monofluoroacetate in Palicourea and Amorimia species.
Toxicon : official journal of the International Society on Toxinology (2012-06-16)
Stephen T Lee, Daniel Cook, Franklin Riet-Correa, James A Pfister, William R Anderson, Flavia G Lima, Dale R Gardner
ZUSAMMENFASSUNG
Numerous plant species worldwide including Palicourea marcgravii and Tanaecium bilabiatum in Brazil cause sudden death and are known to contain monofluoroacetate (MFA). Other species in Brazil including some species traditionally assigned to Mascagnia but now properly called Amorimia species and other Palicourea species are reported to cause sudden death in livestock and are suspected to contain MFA due to the similarity of clinical signs. In this study, an HPLC-APCI-MS method to detect and quantify MFA was developed and was used to investigate plant material from field collections and/or herbarium specimens of Mascagnia, Amorimia, and Palicourea species suspected of causing sudden death. MFA was detected in Amorimia amazonica, Amorimia camporum, Amorimia exotropica, Amorimia pubiflora, Amorimia rigida, and Amorimia septentrionalis as well as Palicourea aeneofusca. MFA concentrations differ greatly between Palicourea species and Amorimia species, which may explain the incidence of poisoning and the amount of plant material required to cause sudden death between these taxa.