Merck
  • Responses of Phanerochaete chrysosporium to toxic pollutants: physiological flux, oxidative stress, and detoxification.
Responses of Phanerochaete chrysosporium to toxic pollutants: physiological flux, oxidative stress, and detoxification.
Environmental science & technology (2012-06-19)
Guang-Ming Zeng, An-Wei Chen, Gui-Qiu Chen, Xin-Jiang Hu, Song Guan, Cui Shang, Lun-Hui Lu, Zheng-Jun Zou
ZUSAMMENFASSUNG
The white-rot fungus Phanerochaete chrysosporium has been widely used for the treatment of waste streams containing heavy metals and toxic organic pollutants. The development of fungal-based treatment technologies requires detailed knowledge of the relationship between bulk water quality and the physiological responses of fungi. A noninvasive microtest technique was used to quantify real-time changes in proton, oxygen, and cadmium ion fluxes following the exposure of P. chrysosporium to environmental toxic (2,4-dichlorophenol and cadmium). Significant changes in H(+) and O(2) flux occurred after exposure to 10 mg/L 2,4-dichlorophenol and 0.1 mM cadmium. Cd(2+) flux decreased with time. Reactive oxygen species formation and antioxidant levels increased after cadmium treatment. Superoxide dismutase activity correlated well with malondialdehyde levels (r(2) = 0.964) at low cadmium concentrations. However, this correlation diminished and malondialdehyde levels significantly increased at the highest cadmium concentration tested. Real-time microscale signatures of H(+), O(2), and Cd(2+) fluxes coupled with oxidative stress analysis can improve our understanding of the physiological responses of P. chrysosporium to toxic pollutants and provide useful information for the development of fungal-based technologies to improve the treatment of wastes cocontaminated with heavy metals and organic pollutants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,4-Dichlorphenol, 99%