Merck
  • Simultaneous determination of baicalin, wogonoside, baicalein, wogonin, oroxylin A and chrysin of Radix scutellariae extract in rat plasma by liquid chromatography tandem mass spectrometry.
Simultaneous determination of baicalin, wogonoside, baicalein, wogonin, oroxylin A and chrysin of Radix scutellariae extract in rat plasma by liquid chromatography tandem mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2012-06-19)
Ling Tong, Meixu Wan, Lihua Zhang, Yonghong Zhu, He Sun, Kaishun Bi
ZUSAMMENFASSUNG
A liquid chromatography tandem mass spectrometry (LC-MS/MS) method was developed for the simultaneous determination of baicalin, wogonoside, baicalein, wogonin, oroxylin A and chrysin in rat plasma, using naringin as an internal standard. After acidifying with HCl, plasma samples were pretreated by liquid-liquid extraction with acetone. Chromatographic separation was accomplished on a Hypersil Gold-C(18) analytical column (2.1×150 mm, 5 μm) utilizing a gradient elution profile and a mobile phase consisting of (A) 0.1% formic acid in water and (B) acetonitrile. Detection was performed by multiple reaction monitoring (MRM) mode using electrospray ionization in the positive ion mode. All analytes showed good linearity over the investigated concentration range (r>0.9900). The lower limit of quantification was 0.5 ng/ml for baicalin, wogonoside, wogonin and oroxylin A, and 1.0 ng/ml for baicalein and chrysin. Intra-day and inter-day precisions (RSD%) were less than 15% and accuracy (RE%) ranged from -6.7% to 5.8%. The validated method was successfully applied to investigate the pharmacokinetics of the major flavonoids of Radix scutellariae extract after oral administration to rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chrysin, ≥96.5%
Supelco
Chrysin, analytical standard
Sigma-Aldrich
Wogonin hydrate, ≥98% (HPLC), solid