Merck
  • Preparation and characterization of uniform particles of flufenamic acid and its calcium and barium salts.
Preparation and characterization of uniform particles of flufenamic acid and its calcium and barium salts.
Journal of colloid and interface science (2012-06-19)
Amr Ali Mohamed, Egon Matijević
ZUSAMMENFASSUNG
Uniform fully dispersed particles of flufenamic acid, a widely used anti-inflammatory drug, were prepared by two different methods. In the first one, the drug solution in organic solvents was added to a non-solvent (water or aqueous solutions of stabilizers); while in the second procedure the drug was precipitated by acidifying its basic aqueous solutions. In addition calcium and barium salts of uniform spherical particles were obtained by precipitation in aqueous basic solutions of the drug. These salts are supposed to improve the drug reactivity. The prepared dispersions of the drug and its salts were examined by scanning electron microscopy, X-ray diffractometry and electrophoresis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
IGEPAL® CA-630, Molecular Biology
Sigma-Aldrich
Bariumchlorid Dihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Bariumchlorid Dihydrat, ≥99%
Sigma-Aldrich
Bariumchlorid, 99.9% trace metals basis
Supelco
IGEPAL® CA-630
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
Hydroxypropylcellulose, average Mw ~80,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropylcellulose, average Mw ~100,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
Bariumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Hydroxypropylcellulose, average Mw ~370,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Triton X-114
Sigma-Aldrich
Triton X-405 -Lösung, 70% in H2O
Sigma-Aldrich
Bariumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hydroxypropylcellulose, average Mw ~1,000,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Bariumchlorid Dihydrat, ≥99.999% trace metals basis
Supelco
Flufenaminsäure, analytical standard, for drug analysis
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
Triton X-165 -Lösung, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Sigma-Aldrich
Bariumchlorid, AnhydroBeads, −10 mesh, 99.95% trace metals basis