- Induction of apoptosis in MCF-7 cells by β-1,3-xylooligosaccharides prepared from Caulerpa lentillifera.
Induction of apoptosis in MCF-7 cells by β-1,3-xylooligosaccharides prepared from Caulerpa lentillifera.
Bioscience, biotechnology, and biochemistry (2012-06-29)
Reiko Maeda, Tomoaki Ida, Hideshi Ihara, Tatsuji Sakamoto
PMID22738982
ZUSAMMENFASSUNG
β-1,3-Xylan was prepared from the green alga, Caulerpa lentillifera, and hydrolyzed to oligosaccharides by a mild acid treatment. The average degree of polymerization was about 5. The oligosaccharides reduced the number of viable human breast cancer MCF-7 cells in a dose-dependent manner, and induced chromatin condensation and degradation of poly ADP-ribose polymerase, indicating that they induced apoptosis in MCF-7 cells.