Merck
  • Renal hemodynamic response to L-arginine in uncomplicated, type 1 diabetes mellitus: the role of buffering anions and tubuloglomerular feedback.
Renal hemodynamic response to L-arginine in uncomplicated, type 1 diabetes mellitus: the role of buffering anions and tubuloglomerular feedback.
American journal of physiology. Renal physiology (2012-06-29)
Alberto Montanari, Almerina Biggi, Aderville Cabassi, Irene Pelloni, Filippo Pigazzani, Silvana Pinelli, Giovanna Pelà, Luisa Musiari, David Z Cherney
ZUSAMMENFASSUNG
According to the "tubulocentric" hypothesis of the glomerular hyperfiltration of diabetes mellitus (DM), tubuloglomerular feedback (TGF) is the critical determinant of the related renal hemodynamic dysfunction. To examine the role of TGF in human type 1 DM, 12 salt-replete healthy (C) and 11 uncomplicated DM individuals underwent measurements of glomerular filtration rate (GFR), renal blood flow (RBF), and lithium-derived absolute "distal" sodium delivery (DDNa). Measurements were made during two 3-h infusions of 0.012 mmol·kg(-1)·min(-1) l-arginine (ARG) buffered with either equimolar HCl (ARG.HCl) or citric acid (ARG.CITR). Our hypothesis was that changes in TGF signaling would be directionally opposite ARG.HCl vs. ARG.CITR according to the effects of the ARG-buffering anion on DDNa. Similar changes in C and DM followed ARG.CITR, with declines in DDNa (-0.26 ± 0.07 mmol/min C vs. -0.31 ± 0.07 mmol/min DM) and increases in RBF (+299 ± 25 vs. +319 ± 29 ml·min(-1)·1.73 m(-2)) and GFR (+6.6 ± 0.8 vs. +11.6 ± 1.2 ml·min(-1)·1.73 m(-2)). In contrast, with ARG.HCl, DDNa rose in both groups (P = 0.001), but the response was 73% greater in DM (+1.50 ± 0.15 mmol/min C vs. +2.59 ± 0.22 mmol/min DM, P = 0.001). RBF also increased (P = 0.001, +219 ± 20 ml·min(-1)·1.73 m(-2) C, +105 ± 14 DM), but ΔRBF after ARG.HCl was lower vs. ARG.CITR in both groups (P = 0.001). After ARG.HCl, ΔRBF also was 50% lower in DM vs. C (P = 0.001) and GFR, unchanged in C, declined in DM (-7.4 ± 0.9 ml·min(-1)·1.73 m(-2), P = 0.02 vs. C). After ARG.HCl, unlike ARG.CITR, DDNa increased in C and DM, associated with less ΔRBF and ΔGFR vs. ARG.CITR. This suggests that the renal hemodynamic response to ARG is influenced substantially by the opposite actions of HCl vs. CITR on DDNa and TGF. In DM, the association of ARG.HCl-induced exaggerated ΔDDNa, blunted ΔRBF, and the decline in GFR vs. C shows an enhanced TGF dependence of renal vasodilatation to ARG, in agreement with a critical role of TGF in DM-related renal hemodynamic dysfunction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Amino-hippursäure, ≥99%