Merck
Antibacterial properties of PES/CuCl(2) three-bore hollow fiber UF membrane.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-07-07)
Jingchuan Dang, Yatao Zhang, Zhan Du, Haoqin Zhang, Jindun Liu
ZUSAMMENFASSUNG
In this study, a three-bore polyethersulfone (PES) hollow fiber ultrafiltration (UF) membrane with antibacterial properties was prepared by phase inversion, using PES as the membrane material, N,N-dimethylacetamide (DMAC) as solvent, polyvinylpyrrolidone (PVP) and CuCl(2) as additives. The effect of CuCl(2) content on the water flux and rejection was studied and the antibacterial properties of PES hollow fiber UF membrane were also investigated. The water flux results indicated that the hydrophilic properties of PES UF membranes were improved after adding CuCl(2). The rejection of PVA-50000 was expected to drop slightly but remain high above 96%. The membranes showed good antibacterial activity against Escherichia coli (E. coli) after adding CuCl(2) and the antibacterial rate of PES/CuCl(2) UF membranes was close to 100% after running for 48 h. PES hollow fiber UF membranes with antibacterial properties were prepared through the formation of the water-soluble PVP/Cu(2+) complex with spatial network structure, which have good antibacterial and hydrophilic properties. Therefore, this study could provide an effective method for membrane antifouling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kupfer(II)-chlorid, 97%
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, reagent grade
Sigma-Aldrich
Kupfer(II)-chlorid, powder, 99%
Sigma-Aldrich
Kupfer(II)-chlorid, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kupfer(II)-chlorid, 99.999% trace metals basis
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, 99.999%
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, ≥99.95% trace metals basis
Sigma-Aldrich
Poly(oxy-1,4-phenylensulfonyl-1,4-phenylen), pellets