Merck
  • Influence of the CO2 absorbent monoethanolamine on growth and carbon fixation by the green alga Scenedesmus sp.
Influence of the CO2 absorbent monoethanolamine on growth and carbon fixation by the green alga Scenedesmus sp.
Bioresource technology (2012-07-10)
Wookjin Choi, Garam Kim, Kisay Lee
ZUSAMMENFASSUNG
The influence of monoethanolamine (MEA) as a CO(2) absorbent on photoautotrophic culture of CO(2)-fixing microalgae was investigated. When 300 ppm MEA (4.92 mM) was added to blank culture medium, the dissolved inorganic carbon and the molar absorption ratio increased to 51.0mg/L and 0.34 mol CO2 = mol MEA, respectively, which was an almost 6-fold increase in CO(2) solubility. When free MEA up to 300 mg/L was added to a green alga Scenedesmus sp. culture that was supplied 5% (v/v) CO(2) at 0.1 vvm, both cell growth rate and final cell density were enhanced compared to when no MEA was added. The cell growth rate reached 288.6 mg/L/d, which was equivalent to 539.6 mg-CO(2)/L/d as a CO(2)-fixation rate and enhancement of about 63.0% compared to not adding MEA. Chlorophyll-a content and nitrate consumption rate increased correspondingly. MEA doses higher than 400mg/L inhibited cell growth, probably due to toxicity of the carbamate intermediate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethanolamin, ≥99%
Sigma-Aldrich
Ethanolamin, ≥98%
Sigma-Aldrich
Ethanolamin, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethanolamin, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamin, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamin, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Ethanolamin, analytical standard