Merck
Amygdalin inhibits angiogenesis in the cultured endothelial cells of diabetic rats.
Indian journal of pathology & microbiology (2012-07-10)
Hossein Mirmiranpour, Shahnaz Khaghani, Ali Zandieh, O Omid Khalilzadeh, Siavash Gerayesh-Nejad, Afsaneh Morteza, Alireza Esteghamati
ZUSAMMENFASSUNG
Angiogenesis contributes to different physiological and pathological conditions. The aim of this study was to investigate for the first time the antiangiogenic effects of amygdalin on the cultured endothelial cells of diabetic rats. A total of 20 streptozotocin-induced diabetic rats were divided into two equal groups of control and amygdalin-treated animals. Eight weeks after the induction of diabetes, amygdalin was injected intraperitoneally (3 mg/kg) to the rats of the treatment group. One day later, rats were sacrificed; the aortic arteries were excised and cut as 2 mm rings. Each aortic ring was incubated in a cell-culture well for 7 days. The process of angiogenesis was monitored by counting the number of microvessels and primary microtubules in each well. Optic microscopy showed proliferation and migration of new endothelial cells to the fibrin gels. The endothelial cells produced primary microtubules which gradually made several branches and finally made a vascular matrix. The number of the primary microtubules and microvessels were significantly lower in the amygdalin-treated vs. control group (P < 0.01). Therefore, amygdalin exerts inhibitory effects on angiogenesis in aortic rings of diabetic rats and may pave a new way for treatment of unfavorable angiogenic conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Amygdalin, ≥99% (HPLC), from apricot kernels
Sigma-Aldrich
Amygdalin, BioXtra, ≥97.0% (HPLC)